Would you like to log in again?
OVEN HI PERFM LOW PWR 220V PRGMBL 300L
Supplier: YAMATO SCIENTIFIC AMERICA
High-performance, energy-saving oven^ High-performance, energy-saving, programmable, constant temperature, forced convection oven, Operating range: RT +10 to 260 deg C, Sensor: K-thermocouple, temp Controller: PID, External dimensions: 730 x 695 x 1685mm, Power source: 220V, Capacity: 300L
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
OVEN HI PERFM LOW PWR 220V PRGMBL 300L 75846-550 DNF-811 Each (300l) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty