Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Economical General Purpose Incubators, Yamato

Supplier: Yamato

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

IC-403CW IC-613CR IC-803CR IC-903CR IC-813CR IC-813CW IC-903CW IC-613CW IC-103CW IC-413CR IC-103C IC-603CR IC-113CW IC-403CR IC-803CW IC-413CW IC-913CW IC-603CW IC-913CR
75846-714EA 1923.06 USD InStock
75846-714 75846-604 75846-614 75846-702 75846-606 75846-716 75846-704 75846-616 75846-608 75846-718 75846-706 75846-708 75846-710 75846-600 75846-610 75846-720 75846-602 75846-712 75846-612
Economical General Purpose Incubators, Yamato
Incubators Compact Benchtop Incubators
The air-jacket technology of these incubators ensures even and efficient heat distribution throughout the chamber.

  • Natural Convection System
  • Operating Temperature Range: Room Temperature +5 to ~80°C
  • Heater Control Circuit: SSR Drive System
  • Temperature Accuracy: ±1.0 °C
  • Temperature Control Accuracy: ±0.5 °C (at 37 °C)
  • Power Outage Compensation
  • Digital PID Controller

The space-saving incubators have a stainless steel interior and a cold-rolled steel plate exterior with a melamine-resin baking finish

The digital setting display and timer are easy to use. A green LED display indicates the measurement temperature, and a red LED display indicates the setting temperature. Both temperatures can be set with keys. Operation functions include fixed temperature, auto start and stop, calibration offset, key lock, and quick auto stop. Safety features include temperature sensor and measurement temperature error and automatic overheat prevention. Self-diagnostic functions make operation safe and error recovery fast.

Benchtop incubators feature stainless steel heating pipes, and larger incubators feature iron-chrome wire heaters. Shelves are provided.

Accessories information: Shelf and bracket set available separately.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty