Would you like to log in again?
Vacuum Drying Oven, Yamato
Supplier: Yamato
ADP-210C ADP-300C ADP-200C ADP-310C
75846-682EA 4040.12 USD InStock
75846-682 75846-684 75846-686 75846-688
Vacuum Drying Oven, Yamato
Ovens Vacuum Ovens
This small benchtop oven uses a decompressed direct heating chamber for vacuum drying.
  • Operating Pressure Range: 101 to ~0.1 kPa (760 to ~1 Torr)
  • Stainless Steel Material
  • Bourdon Tube-Type Vacuum Gauge
  • Mica Heater
  • Temperature Control Accuracy: ±1.5 °C (at 240 °C)
The user-friendly oven is easy to maintain and has straight-forward input of parameters settings and customizable communication ports
The digital PID controller supports fixed temperature, quick auto stop, auto stop and start, calibration offset, and program operations. Each circuit features an independent over-heating prevention device and an over-current electric leakage breaker. Self-diagnostic features guarantee safe operation.
The observational window is made of 12" thick tempered glass and polycarbonate resin plate. A silicone-rubber door seal prevents air from leaking, and the convenient vacuum port is 18 mm dia. The two smaller units weigh approximately 30 kg and heat to full temperate in 70 minutes, and the two larger sizes weigh approximately 55 kg and heat to full temperature in 100 minutes.
Caution: Never use in flammable or explosive gas atmosphere. Never use explosive or flammable material. High-temperature components.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty