Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Large Capacity Laboratory Use Sterilizers, 220-240 V, Yamato

Supplier: Yamato
SQL-810C SQ-510C SQL-1010C SQ-810C
76550-290EA 26176.27 USD InStock
76550-290 75846-690 75846-692 76550-288
Large Capacity Laboratory Use Sterilizers, 220-240 V, Yamato
Sterilizers Autoclaves (Steam Sterilizers)
These ergonomically designed sterilizers are suitable for protein modification and features easy top loading and sample handling.

  • 200-240 V/ 220 V models
  • Axial fan motor
  • Operating temperature range: 45 to ~135 °C
  • Two heaters
  • Three female thread ports for sensors
  • One exhaust valve and one slow-release valve
  • Inner diameter: 370 mm

With simple setting and operation, this sterilizer manages ordinary sterilization, sterilization of culture mediums and liquids, and melting of culture mediums.

The temperature is easy to set and is displayed on a digital screen. Operation modes include an instrument sterilization course, liquid sterilization course, sterilization and keep-warm course, melting course, and custom-programmed course. Custom programs can be saved for repeated use. Safety modes include sensor failure detection, memory-error detection, a lid interlock mechanism, key locking, and a pressure-release valve. The unit can save up to 20 error logs and displays accumulated working and present time.

Units feature an automatic high-pressure steam sterilization system, and the timer start and pre-heating features and cooling fan reduce work time.

Delivery information: Supplied with two mesh baskets.

Caution: 200-240 V/220 V models. Corresponding 50 L model with voltage suitable for use in US (75846-582) is also available, please see separate product page

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty