Would you like to log in again?
Round Metal Specimen Chucks, Suitable for MIROM/QS Series Cryostats, Springside Scientific
Supplier: Springside Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SDAK30OR SDAK40RD SDAK30BK SDAK40GN SDAK30YL SDAK40BL SDAK30BL SDAK40BK SDAK40YL SDAK40OR SDAK30WT SDAK30RD SDAK30PK SDAK40PK SDAK30GN SDAK40WT
75846-770EA 111.78 USD InStock
75846-770 75846-812 75846-768 75846-814 75846-806 75846-816 75846-808 75846-818 75846-760 75846-774 75846-762 75846-776 75846-764 75846-778 75846-810 75846-766
Round Metal Specimen Chucks, Suitable for MIROM/QS Series Cryostats, Springside Scientific
Specimen Mounting Supplies
Round metal specimen chuck with O-ring made of low temperature resistant rubble.
  • Different o-ring colors eliminates sample errors and the use of expensive colored freezing medium
  • Orientation mark to assure proper sample positioning
Proper dimensional tolerance and soft aluminum, to avoid slippage slip in chuck that results in thin/thick cutting.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty