Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Low Density Flat Bags, Elkay Plastics
Supplier: Elkay Plastics
20F-1012 20F-0406 20F-0608 20F-1824 20F-1218 20F-0810 20F-0609 BOR751014
75848-674CS 88.81 USD InStock
75848-674 75848-666 75848-676 75848-670 75848-672 75848-668 75848-678 75848-700
Low Density Flat Bags, Elkay Plastics
Foodservice Supplies
Low Density Polyethylene (LDPE) bags are the most commonly used plastic bag because they provide excellent protection and outstanding value.
  • Made from seamless tubing with no slits or side seals that can rip
  • Bags are 15" wide and under come in dispenser packs of 100 for cleanliness and convenience
  • Cases and inner packs are bar coded and clearly labeled for easy identification
These bags are made from virgin materials, so they comply with FDA requirements.
Certifications: Meet FDA requirements for use in food applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty