Would you like to log in again?
SHAFT SPIN ULTCTR PREC TPR UPPER 16.5IN
Supplier: QLA
Ultracenter Precision Tapered Upper Spin Shaft, 316 Ss, Serialized, For Distek 2100, Size: 16.5^
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SHAFT SPIN ULTCTR PREC TPR UPPER 16.5IN 75849-158 ASTMSHT-16.5 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty