Would you like to log in again?
Twirl’Blue Sterile Sampling Bags, Labplas
Supplier: Labplas
EPR-7012-B EPR-4590-B-VW1
75849-324CS 550.09 USD InStock
75849-324 75849-328
Twirl’Blue Sterile Sampling Bags, Labplas
Bags Sample Bags
These blue sterile sampling bags are specifically conceived for the food industry.
  • Sterile - No cross-contamination
  • Available in different sizes
  • Adapted for all types of samples (solids, semi-solids and liquids)
  • Adapted for all types of homogenisers
  • Economic and efficient way to collect, transport and analyse samples
Collecting samples requires sterile equipment and must be done meticulously to avoid possible sample contaminations. Foreign matter contamination is a possibility in the food industry, therefore Twirl’Blue sterile bags are highly visible in order to work alongside your diligence and avoid possible product recalls.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty