Would you like to log in again?
COLUMN 3% RTX-5 PKD 100/120 6 FT X 2.1MM
Supplier: Avantor
DiatoSorb Diatomaceous Earth Column 3% Rtx-5^ DiatoSorb Diatomaceous Earth Column 3% Rtx-5, (bonded stationary phase), Internal diameter: 2.1 mm, length: 6 ft, Material: Stainless Steel Tubing, Mesh: 100/120 Silcoport W, Packed Column, Low bleed levels, Short conditioning times.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN 3% RTX-5 PKD 100/120 6 FT X 2.1MM 75850-654   Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty