Would you like to log in again?
OIML Class 1 Individual Precision Weights, ASTM, Mettler Toledo
Supplier: Mettler-Toledo
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
11123470 11123474 11123464 11123471 11123446 11123489 11123488 11123470 11123484 11123465 11123490 11123478 11123481 11123489 11123460 11123448 11123487 11123464 11123483 11123477 11123452 11123486 11123478 11123453 11123459 11123463 11123472 11123456 11123469 11123480 11123473 11123491 11123462 11123479 11123467 11123465 11123468 11123450 11123447 11123485 11123471 11123482 11123449 11123475 11123457 11123466 11123451 11123481 11123458 11123473 11123487 11123454 11123476 11123461 11123455
97007-408EA 104 USD InStock
97007-408 97007-448 97007-404 97007-406 97007-444 97007-446 97007-402 97007-452 97007-454 97007-410 CA97007-462 97007-450 89422-918 97007-416 75852-740 CA97007-468 75852-742 97007-418 97007-456 97007-412 97007-414 97007-458 97007-462 75852-738 97007-464 97007-420 97007-460 97007-426 CA97007-438 97007-428 97007-422 97007-466 97007-424 97007-468 97007-474 97007-430 97007-476 97007-432 CA97007-484 CA97007-440 97007-470 97007-472 CA97007-480 97007-438 CA97007-446 CA97007-448 97007-478 97007-434 97007-436 97007-484 97007-440 CA97007-452 97007-442 97007-480 97007-482
OIML Class 1 Individual Precision Weights, ASTM, Mettler Toledo
Weights Calibration Weights
ASTM weights are designed to resist dust and moisture intrusion, offering long-term stability.
  • 316 stainless steel
  • Low magnetic permeability
  • High corrosion resistance
  • High gloss polish
All precision weights are protected in robust and easy to clean plastic boxes. FDA approved materials, including foam inserts, make them a perfect solution for regulated industries.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty