Would you like to log in again?
OIML Class 3 Individual Precision Weights, ASTM, Mettler Toledo
Supplier: Mettler-Toledo
11123559 11123560 11123582 11123583
75852-886EA 2500 USD InStock
75852-886 75852-888 75852-882 75852-884
OIML Class 3 Individual Precision Weights, ASTM, Mettler Toledo
Weights Calibration Weights
ASTM weights are designed to resist dust and moisture intrusion, offering long-term stability.
  • 316 stainless steel
  • Low magnetic permeability
  • High corrosion resistance
  • High gloss polish
  • Stainless steel density: 7.9 kg/dm³
All precision weights are protected in robust and easy to clean plastic boxes. FDA approved materials, including foam inserts, make them a perfect solution for regulated industries.
Experience combined with special skill acquired through years of weight polishing guarantee the consistent high quality our customers demand. Weights are manufactured under stringent ISO 9001 quality management system.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty