Would you like to log in again?
T-Sue™ MicroArray Molds Kits, Simport
Supplier: Simport
M473-170PC M473-24PC M473-15 M473-60PC M473-P30P M473-150PC M473-150 M473-60 M473-24 M473-P40B M473-170 M473-15PC M473-72 M473-36PC M473-72PC M473-36 M473-P10MA M473-P20R M473-P15Y
75853-822PK 882.88 USD InStock
75853-822 75853-998 75853-854 75853-832 75853-996 75853-820 75853-842 75853-830 75853-826 75853-836 75853-824 75853-834 75853-828 75853-838 75854-000 75854-002 75854-004 75853-840 75853-994
T-Sue™ MicroArray Molds Kits, Simport
Microarray Instruments
T-Sue™ Microarray Mold Kits enable tissues from many donor blocks to be arrayed on a single slide.
  • Can be reused hundreds of times without losing its flexibility
  • Withstands temperatures between –100 to 250°C
  • View many different samples on the same slide
The array mold is designed to be simple and easy to use
The precast T-Sue™ Microarray paraffin blocks allow users to prepare tissue arrays without needing any specialized equipment. Construct a tissue array block in minutes, simply by punching the donor tissue cores and inserting them into the pre-made paraffin recipient block. No specific technical training or experience is needed. They are ready to accept cores from a donor block, and offer a consistent core diameter. The cassette is permanently bonded to the block.
T-Sue™ punch needles are used for filling recipient tissue microarray paraffin blocks. Internal stylet operates like a retractable pen. Effortless ejection of core specimens.
Tissues can be analyzed in the same conditions enhancing the efficiency of the research. The T-Sue™ Microarray molds allow users to perform TMAs faster while giving excellent results.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty