Would you like to log in again?
SP Bel-Art H-B Frio-Temp Calibrated Electronic Lollipop Thermometers for Refrigerators, Incubators and General Applications, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
B60210-1800 B60210-2600 B61000-1100 B60210-2200 B60210-1900 B61000-0700 B60210-2100 B60210-1300 B60210-2500 B60210-1700 B61000-0900 B60210-2300 B60210-1400 B61000-0800 B60210-1500 B60210-2700
75854-050CS 395.8 USD InStock
75854-050 75854-052 75854-030 75854-040 75854-054 75854-032 75854-042 75854-034 75854-056 75854-044 75854-036 75854-046 75854-104 75854-038 75854-048 75854-106
SP Bel-Art H-B Frio-Temp Calibrated Electronic Lollipop Thermometers for Refrigerators, Incubators and General Applications, Bel-Art Products, a part of SP
Thermometers Digital Thermometers
Digital verification thermometers verify sample temperatures inside refrigerators and incubators without the use of a cable.
  • Accurate temperature reading without sample contamination is achieved by using the liquid or glass bead filled bottle (included), which mimics the sample
  • All models include accredited calibration documentation that meets the requirements of ISO 17025
  • Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST
  • Individually serialized, CE marked
  • Max./Min.
Includes multi-language instructions (English, French, German, Italian, Portuguese, and Spanish).
Certifications: Meets the requirements of ISO 17025.
Caution: Note: Not for use in ovens or freezers.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty