Would you like to log in again?
Force EXP Guard Column Cartridges, Restek
Supplier: Restek
963450253 963450252 962950252 963950253 962950250 963950252 962950253 963450250 963950250
75854-180PK 557.5 USD InStock
75854-180 75854-228 75854-280 75854-282 75854-230 75854-284 75854-232 75854-176 75854-178
Force EXP Guard Column Cartridges, Restek
Chromatography Columns
Free-Turn architecture lets you change cartridges by hand without breaking inlet/outlet fluid connections, with no tools needed.
  • Titanium hybrid ferrules
  • High pressure seal
  • Auto-adjusting design
Patented titanium hybrid ferrules can be installed repeatedly without compromising high-pressure seal. Auto-adjusting design provides ZDV (zero dead volume) connection to any 10-32 female port. Guard column cartridges require EXP direct connect holder. Pair with EXP hand-tight fitting for tool-free installation. For use with 3 or 5 µm Force LC columns. For 1.8 µm Force columns, use a 0.2 µm UltraShield Filter.
Accessories information: EXP Direct Connect Holder is available separately.
Ordering information: To help protect your investment and further extend the life of our already-rugged LC columns, Restek offers the patent-pending guard column hardware developed by Optimize Technologies. A Restek LC guard cartridge in an EXP direct connect holder is the ultimate in column protection.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty