Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ShelfMax 6" Tall Shelf Bin, Akro-Mils

Supplier: Akro-Mils

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

30048YELLO 30094RED 30048SCLAR 30088BLUE 30088SCLAR 30090SCLAR 30014RED 30098RED 30088RED 30090BLUE 30040RED 30040BLUE 30098BLUE 30040YELLO 30084BLUE 30090RED 30080RED 30014BLUE 30014SCLAR 30048BLUE 30084SCLAR 30094BLUE 30098SCLAR 30080BLUE 30040SCLAR 30094SCLAR 30048RED 30084RED 30080SCLAR 30098YELLO
75855-768CS 372.82 USD InStock
75855-768 75854-740 75855-908 75854-760 75854-710 75854-732 75854-754 75854-730 75854-758 75854-714 75854-712 75854-734 75854-756 75854-718 75854-738 75854-716 75855-770 75854-692 75854-694 75854-750 75854-696 75854-722 75854-686 75854-742 75854-748 75854-726 75854-746 75854-724 75854-708 75854-706
ShelfMax 6" Tall Shelf Bin, Akro-Mils
Boxes Dispenser Boxes
Industry-leading, heavy-duty ShelfMax bins offer the best value in 6" high plastic storage bins.

  • Reinforced side ribs for added strength
  • Built-in rear hanglock allows bin to tilt out for complete access
  • Made in the USA

The 6" tall bins feature greater storage options for end users, including up to 85% more holding capacity than traditional 4" shelf bins. ShelfMax bins are a superior alternative to weak and flimsy cardboard bins and multiple divider slots allow for a greater number of storage options. Bins work with standard shelving, or vertical storage and retrieval units. One-piece construction is waterproof and unaffected by grease or oil.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty