AkroBins are the industry-leading hanging and stacking plastic storage bin.
- Anti-slide stop prevents stacked bins from shifting forward
- Full-width hanger lip supports bin on hanging system
- Made in the USA
Heavy-duty, industrial-grade polymer bins stack together or hang from Akro-Mils louvered racks and panels, rail racks, storage cabinets, and louvered carts for high-density storage. AkroBins are corrosion-proof and unaffected by weak acids and alkalis. Dividers, lids, and a wide range of labeling options are available separately.
Accessories information: Create multiple compartments within a single bin, and keep items separated with economical and sturdy length dividers. This provides additional storage options, to maximize space and efficiency.