Would you like to log in again?
Akro-Grid Dividable Boxes, Akro-Mils
Supplier: Akro-Mils
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Akro-Grid
33223GREY 33168GREY 33224RED 33224BLUE 33220BLUE 33228GREY 33226GREY 33228RED 33164RED 33105GREY 33162RED 33166RED 33220RED 33166GREY 33224GREY 33105RED 33220GREY 33164GREY 33105BLUE 33223RED 33223BLUE 33166BLUE 33226RED 33226BLUE 33168BLUE 33164BLUE
75855-466CS 278.95 USD InStock
75855-466 75855-444 75855-488 75855-422 75855-402 75855-468 75855-448 75855-404 75855-406 75855-490 75855-472 75855-450 75855-452 75855-474 75855-476 75855-434 75855-414 75855-436 75855-438 75855-418 75855-480 75855-482 75855-484 75855-420 75855-464 75855-442
Akro-Grid Dividable Boxes, Akro-Mils
Boxes Storage Boxes
Akro-Grid Dividable Boxes are ideal for work-in-process transfer applications
  • Divider slots allow users to create hundreds of compartment sizes
  • Dividers (sold separately) allow subdivision of the containers to a 1.125" square compartment size
  • Made in the USA
Boxes resist most solvents and chemicals. Strong stacking rims and external ribbing provide added strength. Optional snap-on lids keep contents secure and dust free. Dividers are sold separately.
Use dividers to create custom sized storage compartments as small as 1.125".
Accessories information: Optional Accessory include Clip-On Tote Label Holder clip on to Akro-Grid Dividable Containers to provide a highly visible label for parts tracking and barcode scanning.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty