Would you like to log in again?
Lysing Matrix Z, MP Biomedicals
Supplier: MP Biomedicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
116961500 116961050 116961050 116985025 116540438 116540439 116978025 116978050 116985100 116540439 116978005 116978005 116540438 116978025 116978050 116985100 116985050 116985025 116985050 116961500
10015-622PK 541.45 USD InStock
CA76045-762 CA76045-760 10015-622 75856-408 CA76045-764 CA76045-786 CA76046-228 75856-410 CA76046-078 CA76046-088 75856-386 75856-166 75856-130 75856-164 75856-140 75856-162 10015-624 CA76046-052 CA76046-054 CA76046-230
Lysing Matrix Z, MP Biomedicals
Protein Extraction Protein Extraction Reagents
Lysing Matrix Z is used for breaking tough plant and animal samples.
  • High density and high hardness
  • Ideal for cracking very tough cell walls
  • Smooth surface minimizes shearing
  • Lysis of gram positive and gram negative bacteria, fungal tissue and spores, yeasts, molds, filamentous algaes and biofilms
Lysing Matrices are critical components of the FastPrep™ Sample Preparation system. MP Biomedicals recieves the matrix particles from proven vendors and ensures the unprocessed material meets high qualitative standards. Matrix particles are then prepared and dispensed under a rigorous set of proprietary conditions that offer customers a premium product that can be immediately used with confidence. Matrices are available separately for use with your own unique buffers, and are also available as components of the complete purification kits.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty