Would you like to log in again?
SmartBlocks for ThermoMixer® C and ThermoStat C
Supplier: Eppendorf
5318000001 5318000001 5362000035 5361000031 5362000035 5310000002 5364000024 5363000039 5361000031 5365000028 5306000006 5366000021 5307000000 5306000006 5364000024 5309000007 5367000025 5316000004 5309000007 5360000038 5307000000 5366000021 5316000004 5360000038 5310000002 5367000025 5365000028 5363000039
89428-672EA 1140.18 USD InStock
CA11028-286 89428-672 CA11028-284 89428-674 CA11028-300 CA11028-288 89428-670 75856-524 CA11028-282 75856-522 89428-668 89428-664 89428-666 89428-682 CA11028-296 89428-684 CA11028-294 89428-680 CA11028-298 CA75856-524 CA75856-522 CA77321-626 CA11028-292 CA11028-290 77321-626 CA11028-302 89428-676 89428-678
SmartBlocks for ThermoMixer® C and ThermoStat C
Block Heaters Blocks
Exchangeable in just seconds. Individually sensor-controlled SmartBlocks are your solution for maintaining accurate, uniform temperatures. A wide variety of these flexible thermoblocks for all common laboratory vessels support a broad range of applications - for use with Eppendorf ThermoMixer® C and ThermoStat C.
  • Optimized block design supports optimal temperature transfer to sample to achieve excellent results
  • Eppendorf SmartBlock-specific calibration by integrated sensor technology maximizes temperature accuracy and homogeneity
  • Insulated thermoblocks for ergonomic operation - you won't burn your fingers (PhysioCare Concept®)
  • Automatic block recognition for safe handling
  • Optimized block design ensures maximum temperature transfer to the sample
  • Eppendorf QuickRelease technology enables fast and simple block exchange – no need for tools
Every Eppendorf SmartBlock is individually inspected in production for temperature accuracy and includes a serial-number specific certificate for documentation purpose.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty