Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

VWR® 35 mm Confocal Dish, Sterile

Supplier: VWR
VWR®
BDD012035 BDD002035 BDD013035 BDD003035
75856-746CS 701.45 USD InStock
75856-746 75856-744 75856-742 75856-740
VWR® 35 mm Confocal Dish, Sterile
Culture Dishes Cell Culture Dishes
Dish surfaces are smooth and free from striation to maximize usable area for growth. The rim on upper side of the lid mates with the dish brim for easy and secure stacking. Dish lids feature several small chimbs to shape vents, and are positioned for effective gas exchange. Sterilized by irradiation. Non-pyrogenic.

  • Available with 2 different glass bottom sizes of 15mm or 20mm
  • Borosilicate glass construction, with high homogeneity, low bubble, and low inclusion content
  • Glass thickness is 0.16 mm
  • Medical grade shadowless glue, highly transparent achromatic
  • Suitable for all for all living cell examinations

Certifications: ISO13485: 2003 and ISO9001: 2008.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty