Would you like to log in again?
KIMAX® Reusable Volumetric Pipettes, Class B, Color-Coded, Kimble Chase
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KIMAX®
37001-3 37001-20 37001-10 37001-1 37001-25 37001-50 37001-100 37001-2 37001-5
75856-770CS 105.81 USD InStock
75856-770 75856-782 75856-780 75856-778 75856-776 75856-774 75856-786 75856-772 75856-784
KIMAX® Reusable Volumetric Pipettes, Class B, Color-Coded, Kimble Chase
Pipettes Graduated Pipettes
These rugged volumetric pipettes are calibrated to deliver and feature a legend of permanent brown stain.
  • Large numerals on the bulb indicate capacity
  • Color-coding (ASTM E1273) facilitates sorting and selection
Designed from ASTM Specification E969, Class B requirements.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty