Would you like to log in again?
Saline 0.9%, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
U264 U264
75860-368PK 270.58 USD InStock
75860-368 CA75870-682
Saline 0.9%, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
Saline 0.9% for sterility testing filter rinsing.
  • Sterility testing filter rinse
Ordering information: Minimum Order of 28 Packs.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty