Would you like to log in again?
Combined Titration Electrodes, Plug and Play, Mettler
Supplier: Mettler-Toledo
51109531 51109530 51109520
75870-680EA 620 USD InStock
75870-680 75870-676 75870-678
Combined Titration Electrodes, Plug and Play, Mettler
Electrodes pH Electrodes
Plug & Play combined silver ring electrode for argentometric titrations.
  • Plug & Play sensor with integrated sensor chip for secure handling
  • Sensor ship stores data such as sensor type, serial number, calibration data and usable life, which is automatically read out to the titrator setup when connected
  • Ceramic frit junction
  • Pure silver sensing element
  • ARGENTHAL™ reference system
  • Refillable reference electrolyte (1 mol/L KNO3)
Typical applications include: Titrations of halides, sulfides, hydrogen sulfide, mercaptans and cyanides in aqueous medium.
Examples: Salt content (rock salt, NaCl) in ketchup, food, vegetable juices; sulphide content of green and black liquor in the pulp and paper industry; silver content of silver alloys.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty