Would you like to log in again?
CELLdisc™ MultiLayer Cell Culture Systems, Greiner Bio-one
Supplier: Greiner bio-one
878072 678940 678140 678104 678916 678904 878075 678908 678108 678116 878071 678101 878074
76311-054CS 2299.65 USD InStock
76311-054 75870-804 76311-052 75870-758 76311-050 75870-808 76457-316 75870-806 76311-046 75870-760 75870-764 75870-762 75870-766
CELLdisc™ MultiLayer Cell Culture Systems, Greiner Bio-one
Bioreactors
CELLdisc™ is a multilayer device covering a range of cell culture surfaces from 250 cm2 up to 10000 cm2. The innovative, ergonomic design provides a versatile system from research scale to industrial batches.
  • Non-pyrogenic, non-cytotoxic
  • Free from DNase, RNase, and human DNA
  • Screw cap color indicating surface treatment
  • Integrated gas support channel and vent port for pressure equilibration
  • Available in 4-, 8-, 16-, and 40-layer versions
A centrally located channel allows uniform distribution of gas throughout the device. A socle-rim guarantees that the bottom layer of the CELLdisc™ does not touch the surface of the incubator. These compact and robust cylindrical devices are ideally suited for automation and upscaling of mass cell culture. A large 38mm opening with screw cap enables easy filling.
Certifications: USP Class VI and Sterility Assurance Level (SAL) of 10–6.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty