Would you like to log in again?
SafTest Dispenser Sets, MP Biomedicals, LLC
Supplier: MP Biomedicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
07DSFA2000 07DSTG2000 07DSAK2000 07DSPF2000 07DSPR2000 07DSMA2000
75870-868EA 2145.56 USD InStock
75870-868 75870-870 75870-874 75870-872 75870-866 75870-876
SafTest Dispenser Sets, MP Biomedicals, LLC
Bottle Top Dispensers
Pre-set volumetric dispensers which allow for quick analysis.
  • Choose the dispenser set for each chosen endpoint
  • Allows for accurate testing, every time
  • For use with SafTest kits
Peroxide Test Kit Dispenser Set includes one dispenser for each Reagent (A, B, C, and Preparation Reagent) and one reagent holding rack.
Free Fatty Acid Test Kit Dispenser Set includes one dispenser for each Reagent (A, B, and Preparation Reagent) and one reagent holding rack.
Malonaldehyde Test Kit Dispenser Set includes one dispenser for each Reagent (A, B, and Preparation Reagent) and one reagent holding rack.
Alkenal Test Kit Dispenser Set includes one dispenser for each Reagent (A, B, and Preparation Reagent) and one reagent holding rack.
Total Glycerin Test Kit Dispenser Set includes one dispenser for each Reagent (A and Preparation Reagent) and one reagent holding rack.
Caution: Storage: Room temperature
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty