Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Aerosolv® Recycling System and Components, Justrite®
Supplier: Justrite
Aerosolv®
28231 28223 28224 28229 28225 28228 28230
75870-990EA 304.94 USD InStock
75870-990 75871-154 75871-156 75870-994 75870-992 75870-988 75870-986
Aerosolv® Recycling System and Components, Justrite®
Safety Cans
Safely recycle aerosol cans while reducing disposal costs and minimizing waste.
  • Save thousands of dollars on hazardous waste disposal costs
  • Turn aerosolv cans into recyclable steel; save on landfill space for a cleaner environment
  • The only system certified by the United States Environmental Protection Agency
The Aerosolv® System safely converts spent aerosol cans from solid hazardous waste to a non-hazardous material
Attach the two-part combination filter to the 0.75" bung to filter VOCs from any residual propellants. The bottom coalescing cartridge transforms residual liquids into droplets that collect in the reservoir of the cartridge and safely drain into the drum. The top carbon filter cartridge indicates saturation and serves as a replacement reminder.
It accepts industry standard sized cans with a maximum height of 8.5", and threads directly into a standard 2" bung on a 30 or 55 gallon drum. The body is made of low maintenance aluminum and all moving parts are 308 stainless steel. A non-sparking pin punctures the can, and the residual liquids flow into a collection drum. One 55 gallon drum holds approximately 4,200 spent aerosol cans. Punctured, discharged cans do not meet the definition of hazardous waste per EPA 40 CFR 261.23(a)(6), and thus can be treated as recyclable scrap metal.
Certifications: EPA Compliant and ETV certified. The Aerosolv® Dual Compliant Can Disposal System with Counter is California Compliant as well.
Accessories information: Full systems include the puncturing unit, a combination coalescing/saturation indicating carbon filter, a padlock with key, antistatic ground wire with C-clamp, and safety goggles. Replacement and accessory components are available separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty