Would you like to log in again?
DRG® TGF-ß1 ELISA, DRG International
Supplier: DRG International
DRG®
EIA-1864
75871-070KT 369.96 USD InStock
75871-070
DRG® TGF-ß1 ELISA, DRG International
Assays ELISAs
An enzyme immunoassay for the quantitative measurement of TGF‑β1 in serum and cell culture supernatant.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
Transforming Growth Factor β1 (TGF-β1) is a 25 kDa Homodimer composed of two 12.5 kDa subunits joined by disulfide bonds
The DRG TGF‑β1 ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. Prior to testing the standards and samples are diluted in assay buffer, acidified with HCl and then neutralized with Neutralization Buffer. Thereafter, the neutralized standards and samples are added to the antibody coated (polyclonal) microtiter wells. After incubation unbound sample material is removed by washing. In a second step monoclonal mouse anti TGF-β1 antibody, a biotinylated anti mouse IgG antibody and the Streptavidin-HRP Enzyme complex are incubated successively, forming an immuno enzyme sandwich complex. After incubation the unbound conjugate is washed off. Having added the substrate solution, the intensity of color developed is proportional to the concentration of TGF β1 in the sample.
TGF-β1 is a multipotent Cytokine with cell- and dose-dependent activities. This molecule is produced by a number of cells and tissue types, e.g. thrombocytes, bone tissue, placenta and kidneys. This potent Cytokine modulates embryonic development, bone formation, mammary development, wound healing, hematopoiesis, cell cycle progression and the production of the extracellular matrix. With respect to the immune system, TGF-β1 inhibits T and B cell proliferation and acts as an anti-inflammatory molecule both in vitro and in vivo. TGF-β1 inhibits macrophage maturation and activation. This molecule also inhibits the activity of natural killer cells and lymphokine activated killer cells and blocks cytokine production.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty