Would you like to log in again?
DRG® TGF-β2 ELISA, DRG International
Supplier: DRG International
DRG®
EIA-2396
75871-074KT 325.13 USD InStock
75871-074
DRG® TGF-β2 ELISA, DRG International
Assays ELISAs
An enzyme immunoassay for the quantitative measurement of transforming growth factor β2 (TGF‑β2) in serum, plasma, and cell culture supernatant.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
Transforming growth factor-β (TGF‑β) is a multipotent Cytokine with cell- and dose-dependent activities
The DRG TGF‑β2 ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. Prior to testing the standards and patient samples are diluted in assay buffer, acidified with HCl and then neutralized with NaOH. Afterwards, the neutralized standards and samples are added to the antibody coated (polyclonal) microtiter wells. After the first incubation the unbound sample material is removed by washing. Then a biotinilated mouse anti TGF-β2 antibody and the Streptavidin-HRP Enzyme complex are incubated in succession. An immuno enzyme sandwich complex is formed. After incubation the unbound conjugate is washed off. Having added the substrate solution, the intensity of colour developed is proportional to the concentration of TGF‑β2 in the patient sample.
This molecule is produced by a number of cells and tissue types, e.g. thrombocytes, bone tissue, placenta and kidneys. TGF-β2 is not produced by blood platelets, contrary to TGF‑β1. This potent Cytokine modulates embryonic development, bone formation, mammary development, wound healing, hematopoiesis, cell cycle progression and the production of the extracellcular matrix. TGF‑β2– null mice were shown to exhibit perinatal mortality and a wide range of developmental defects for a single gene description which include cardiac, lung, craniofacial, limb, spinal column, eye, inner ear and urogenitial defects. TGF‑β2 has been shown to be a potent growth inhibit factor in the regulation of postnatal cerebellar neurons and neuroblast proliferation. TGF‑β2 has been detected in tear fluid. TGF‑β2 levels are elevated in the vitreous of patients with proliferative diabetic retinopathy. Elevated plasma levels of TGF‑β2 have been described in patients with disseminated malignant melanoma. TGF‑β2 concentrations are furthermore elevated in Parkinson’s disease in ventricular cerebrospinal fluid.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty