Would you like to log in again?
DRG® Corticosterone ELISA, DRG International, Inc.
Supplier: DRG International
DRG®
EIA-4164
75871-076KT 343.6 USD InStock
75871-076
DRG® Corticosterone ELISA, DRG International, Inc.
Assays ELISAs
An enzyme immunoassay for the quantitative measurement of corticosterone in serum and plasma.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
Corticosterone is a glucocorticoid secreted by the cortex of the adrenal gland
The DRG Corticosterone ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. The microtiter wells are coated with a polyclonal antibody directed towards an antigenic site on the Corticosterone molecule. Endogenous Corticosterone of a patient sample competes with a Corticosterone-horseradish peroxidase conjugate for binding to the coated antibody. After incubation the unbound conjugate is washed off. The amount of bound peroxidase conjugate is reverse proportional to the concentration of Corticosterone in the sample. After addition of the substrate solution, the intensity of colour developed is reverse proportional to the concentration of Corticosterone in the patient sample.
Corticosterone is produced in response to the stimulation of the adrenal cortex by adrenocorticotropic hormone (ACTH) and is the precursor of aldosterone. Corticosterone is a major indicator of stress since stress increases the production of corticosteroids. Studies involving corticosterone and levels of stress include impairment of long term memory retrieval1, chronic corticosterone elevation due to dietary restrictions and in response to burn injuries. In addition to stress levels, corticosterone is believed to play a decisive role in sleep-wake patterns.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty