Would you like to log in again?
DRG® Leptin Sandwich ELISA, DRG International
Supplier: DRG International
DRG®
EIA-2395R
75871-088KT 225.3 USD InStock
75871-088
DRG® Leptin Sandwich ELISA, DRG International
Assays ELISAs
An enzyme immunoassay for the quantitative measurement of leptin in serum and plasma.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
The DRG Leptin ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal antibody directed towards a unique antigenic site on a Leptin molecule. An aliquot of patient sample containing endogenous Leptin is incubated in the coated well with a specific biotinylated monoclonal anti Leptin antibody. A sandwich complex is formed. After incubation the unbound material is washed off and a Steptavidin Peroxidase Enzyme Complex is added for detection of the bound Leptin. Having added the substrate solution, the intensity of colour developed is proportional to the concentration of Leptin in the patient sample.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty