Would you like to log in again?
DRG® T4 Total ELISA, DRG International
Supplier: DRG International
DRG®
EIA-4568
75871-090KT 107.95 USD InStock
75871-090
DRG® T4 Total ELISA, DRG International
Assays ELISAs
An enzyme immunoassay for the quantitative measurement of total thyroxin (T4) in serum or plasma.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
Measurement of serum thyroxine concentration is generally regarded as an important in-vitro diagnostic test for assessing thyroid function
The DRG Total T4 ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. The microtiter wells are coated with a monoclonal [mouse] antibody directed towards an antigenic site of the T4 molecule. Endogenous T4 of a patient sample competes with a T4-horseradish peroxidase conjugate for binding to the coated antibody. After incubation the unbound conjugate is washed off. The amount of bound peroxidase conjugate is inversely proportional to the concentration of T4 in the sample. After addition of the substrate solution, the intensity of colour developed is inversely proportional to the concentration of T4 in the patient sample.
This importance has provided the impetus for the significant improvement in assay methodology that has occurred in the last three decades. This procedural evolution can be traced from the empirical protein bound iodine (PBI) test to the theoretically sophisticated radioimmunoassay. Not intended for newborn screening.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty