Would you like to log in again?
DRG® Bordetella pertussis IgG ELISA, DRG International
Supplier: DRG International
DRG®
EIA-3450
75871-096KT 171.12 USD InStock
75871-096
DRG® Bordetella pertussis IgG ELISA, DRG International
Assays ELISAs
An enzyme immunoassay for the qualitative and semiquantitative determination of IgG-class antibodies to Bordetella pertussis and Bordetella pertussis toxin in serum.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
Bordetella species are non-spore-forming encapsulated bipolar, coccoid (pale-staining) Gram-negative bacilli (about 0.3 to 0.5 µm thick and 1 µm long)
The DRG Bordetella pertussis/toxin IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Microtiter wells as a solid phase are coated with Bordetella pertussis and Bordetella pertussis toxin antigen. Diluted patient specimens and ready-for-use controls are pipetted into these wells. During incubation Bordetella pertussis and Bordetella pertussis toxin-specific antibodies of positive specimens and controls are bound to the immobilized antigens. After a washing step to remove unbound sample and control material horseradish peroxidase conjugated anti-human IgG antibodies are dispensed into the wells. During a second incubation this anti‑IgG conjugate binds specifically to IgG antibodies resulting in the formation of enzyme-linked immune complexes. After a second washing step to remove unbound conjugate the immune complexes formed (in case of positive results) are detected by incubation with TMB substrate and development of a blue color. The blue color turns into yellow by stopping the enzymatic indicator reaction with sulfuric acid. The intensity of this color is directly proportional to the amount of Bordetella pertussis and Bordetella pertussis toxin-specific IgG antibody in the patient specimen. Absorbance at 450 nm is read using an ELISA microtiter plate reader.
The genus consists of the human parasites B. pertussis and B. parapertussis, and B. bronchiseptica which cause enzootic infections in various wild and domestic animal species. Bordetella pertussis produces a single-disease syndrome in man known as pertussis or whooping cough. It is a highly contagious childhood disease (approx. 80% of cases occur before the age of five) which is transmitted by respiratory contact and is associated with a high mortality rate (about 1 to 2% in the first year of life, later on about 1%). In the absence of immunization, essentially no one escapes pertussis. Clinical pertussis is followed by natural acquired immunity which is long-lasting but not permanent. The distribution of the disease is worldwide, though clearly modified by immunization and other poorly defined social, economic, and nutritional factors. In most countries an active vaccination is recommended. Usually the immunization preparation is combined with diphtheria and tetanus toxoids.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty