Would you like to log in again?
DRG® PMSG ELISA, DRG International
Supplier: DRG International
DRG®
EIA-1298
75871-100KT 470 USD InStock
75871-100
DRG® PMSG ELISA, DRG International
Assays ELISAs
An enzyme immunoassay for the quantitative measurement of pregnant mare serum gonadotropin (PMSG) in equine serum and plasma.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
PMSG or equine chorionic gonadotropin is secreted by the endometrial cups of the pregnant mares uterus. The hormone is found in the blood of the pregnant mare between days 40 to 120 of gestation, reaching a peak at approximately day 60. Measurement of PMSG provides a specific test for pregnancy since the hormone is only found in the pregnant mare. After 150 days of gestation, hormone levels are no longer detectable. The PMSG diagnostic test as a means of pregnancy testing is a suitable and convenient method to confirm earlier ultrasound scanning techniques and allows the practitioner to obtain this information quickly and economically in the laboratory. The DRG PMSG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal antibody directed towards a unique antigenic site of the PMSG molecule. An aliquot of sample containing endogenous PMSG is incubated in the coated well. After washing a second incubation follows with enzyme conjugate, which is an anti-PMSG antibody conjugated with horseradish peroxidase. After incubation the unbound conjugate is washed off. The amount of bound peroxidase is proportional to the concentration of PMSG in the sample. Having added the substrate solution, the intensity of color developed is proportional to the concentration of PMSG in the sample.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty