Would you like to log in again?
DRG® TM-CA 19-9 ELISA, DRG International
Supplier: DRG International
DRG®
EIA-5069R
75871-106KT 283.8 USD InStock
75871-106
DRG® TM-CA 19-9 ELISA, DRG International
Assays ELISAs
An enzyme immunoassay for the quantitative measurement of CA 19‑9 in serum or plasma.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
The sialyl Lewisaepitope is also known as the cancer associated antigen CA 19-9. CA 19-9 levels are frequently elevated in the serum of patients with cancers of the pancreato-biliary system (i.e. pancreas, gallbladder, biliary tract). In addition, elevated levels of CA 19-9 have been observed in other malignancies such as lung cancer, other gastrointestinal cancers and in some nonmalignant disorders. It must be taken into acount that patients, genotypically negative for the Lewis blood group antigen, will be unable to produce the CA 19-9 antigen even in the presence of malignant tissue. The DRG TM‑CA 19‑9 ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal [mouse] antibody directed towards a unique antigenic site of the CA 19‑9 molecule. An aliquot of patient sample containing endogenous CA 19‑9 is incubated in the coated well with assay buffer. After a washing step a second incubation follows with enzyme conjugate, which is an anti-CA 19‑9 antibody conjugated with horseradish peroxidase. After incubation the unbound conjugate is washed off. The amount of bound peroxidase is proportional to the concentration of CA 19‑9 in the sample. Having added the substrate solution, the intensity of colour developed is proportional to the concentration of CA 19‑9 in the patient sample.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty