Would you like to log in again?
DRG® 17-OH Progesterone ELISA, DRG International, Inc.
Supplier: DRG International
DRG®
EIA-1292
75871-116KT 183.96 USD InStock
75871-116
DRG® 17-OH Progesterone ELISA, DRG International, Inc.
Assays ELISAs
An enzyme immunoassay for the quantitative measurement of 17-α-OH progesterone (17-α-OHP) in serum.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
The steroid 17-α-Hydroxyprogesterone (17-α-OHP) is produced by both the adrenal cortex and gonads
The DRG 17-OH Progesterone ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. The microtiter wells are coated with a polyclonal [goat] antibody directed towards an antigenic site of the 17‑α‑OHP molecule. Endogenous 17-α-OHP of a sample competes with a 17-α-OHP-horseradish peroxidase conjugate for binding to the coated antibody. After incubation the unbound conjugate is washed off.
The amount of bound peroxidase conjugate is inversely proportional to the concentration of 17-α-OHP in the sample. After addition of the substrate solution, the intensity of color developed is inversely proportional to the concentration of 17-α-OHP in the sample.
Even though 17-α-OHP has relatively little progestational activity, it is of intense clinical interest because it is the immediate precursor to 11-desoxycortisol (Cpd-S). Because Cpd-S is produced by 21-hydroxylation of 17-α-OHP, measurement of 17-α-OHP is a useful indirect indicator of 21-hydroxylase activity. In congenital 21-hydroxylase deficiency, the most common variety of congenital adrenal hyperplasia (CAH), 17-α-OHP is secreted in abundant excess. It is moderately elevated in the 11-β-hydroxylase deficiency as well. Measurement of 17-α-OHP is therefore valuable in the initial diagnosis of CAH.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty