Would you like to log in again?
DRG® Progesterone ELISA, DRG International
Supplier: DRG International
DRG®
EIA-1561
75871-122KT 186.65 USD InStock
75871-122
DRG® Progesterone ELISA, DRG International
Assays ELISAs
An enzyme immunoassay for the quantitative determination of progesterone in serum and plasma.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
Progesterone (pregn-4-ene-3, 20-dione) is a C21 steroid hormone containing a keto-group (at C-3) and a double bond between C-4 and C-5 (Δ4)
The DRG Progesterone ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. The microtiter wells are coated with a polyclonal antibody directed towards an antigenic site on the Progesterone molecule. Endogenous Progesterone of a sample competes with a Progesterone horseradish peroxidase conjugate for binding to the coated antibody. After incubation the unbound conjugate is washed off. The amount of bound peroxidase conjugate is reverse proportional to the concentration of Progesterone in the sample. After addition of the substrate solution, the intensity of color developed is reverse proportional to the concentration of Progesterone in the sample.
This steroid hormone is a female sex hormone which, in conjunction with estrogens, regulates the accessory organs during the menstrual cycle and it is particularly important in preparing the endometrium for the implantation of the blastocyte and in maintaining pregnancy. In non-pregnant women progesterone is mainly secreted by the corpus luteum whereas in pregnancy the placenta becomes the major source. Minor sources are the adrenal cortex for both sexes and the testes for males. Progesterone circulates in blood mainly bound to Corticosteroid Binding Globulin (CBG), Sex Hormone Binding Globulin (SHBG) and Albumin. Only 2-10% of the total concentration circulates as free hormone. Blood progesterone concentrations vary widely according to the phases of menstrual cycle; they are lower than 1 ng/mL (3.2 nmol/L) in the follicular phase and around 10 to 20 ng/mL (32 to 64 nmol/L) in the luteal phase. The maximal levels are achieved 4 to 7 days after ovulation and remain elevated for 4 to 6 additional days prior to falling to the preovulatory levels 24 hours before the onset of menstruation.Since the rise and fall of progesterone parallel the activity of ovarian follicle and corpus luteum, measurements of plasma progesterone are clinically used to confirm ovulation and normal function of the corpus luteum in non-pregnant women. If ovulation does not occur the corpus luteum is not formed and no cyclical rise of progesterone in plasma is observed. Abnormal progesterone secretion has been implicated in premenstrual tension, irregular shedding of endometrium, dysmenorrhoea, and luteal insufficiency. Progesterone concentration can vary not only from subject to subject but also in the same person from day to day or even from hour to hour. Consequently, in gynecological disorders or abnormal pregnancies serial measurements rather than single ones are recommended for a proper interpretation of results. During pregnancy progesterone is widely produced by placenta, and plasma levels rise steadily achieving values as high as 200 ng/mL at term.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty