Would you like to log in again?
DRG® DHEA-S ELISA, DRG International
Supplier: DRG International
DRG®
EIA-1562
75871-124KT 183.96 USD InStock
75871-124
DRG® DHEA-S ELISA, DRG International
Assays ELISAs
An enzyme immunoassay for the quantitative measurement of DHEA‑S in serum and plasma.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
Dehydroepiandrosterone (5-Androstene-3β-0L-17-one, Androstenolone, Dehydroisoandrosterone, Transdehydroandrosterone, DHEA) is a steroid hormone present in blood mostly in its sulfate form (DHEA S)
The DRG DHEA‑S ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. The microtiter wells are coated with a polyclonal antibody directed towards an antigenic site of the DHEA‑S molecule. Endogenous DHEA‑S of a patient sample competes with a DHEA‑S-horseradish peroxidase conjugate for binding to the coated antibody. After incubation the unbound conjugate is washed off. The amount of bound peroxidase conjugate is inversely proportional to the concentration of DHEA‑S in the sample. After addition of the substrate solution, the intensity of colour developed is inversely proportional to the concentration of DHEA‑S in the patient sample.
DHEA-S is a more specific product of the adrenals and measurements of this steroid are widely used in clinical practice. The clinical importance of plasma assays of DHEA-S is associated with the diagnosis of adrenal hyperplasia and differential diagnosis of hirsutism.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty