Would you like to log in again?
DRG® Immunoglobulin E (IgE) ELISA, DRG International
Supplier: DRG International
DRG®
EIA-1788
75871-130KT 142.61 USD InStock
75871-130
DRG® Immunoglobulin E (IgE) ELISA, DRG International
Assays ELISAs
An enzyme immunoassay for the quantitative determination of immunoglobulin E (IgE) concentration in serum.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
Patients with atopic allergic diseases such as atopic asthma, atopic dermatitis, and hay fever have been shown to exhibit increased total Immunoglobulin E (IgE) levels in blood
Decreased levels of IgE are found in cases of hypogammaglobulinemia, autoimmune diseases, ulcerative colitis, hepatitis, cancer, and malaria.6 Cord blood or serum IgE levels may have prognostic value in assessing the risk of future allergic conditions in children.
Certain groups of white blood cells, including basophils and tissue mast cells, have membrane receptors for the IgE molecule. These target cells, through a series of complex reactions, form a combination of a specific allergen with antibody-sensitized basophils or mast cells, and initiate the release of certain vasoactive agents, such as histamine, into the blood stream.
As a result, there is a constriction of smooth muscles, dilation of small blood vessels, activation of blood platelets, and irritation of skin nerve endings characteristic of allergic reactions. Typical clinical symptoms of immediate hypersensitivity are inflammation and itching in a skin reaction, or congestion in a bronchial reaction. The IgE serum concentration in a patient is dependent on both the extent of the allergic reaction and the number of different allergens to which the patient is sensitized. Non-allergic normal individuals have IgE concentrations that vary widely and increase steadily during childhood, reaching their highest levels at age 15 to 20, and thereafter remaining constant until about age 60, when they slowly declineThe DRG IgE Quantitative Test is a solid phase enzyme¬linked immunosorbent assay (ELISA) based on the sandwich principle.
IgE is also known as the reaginic antibody. In general, elevated levels of IgE indicate an increased probability of an IgE-mediated hypersensitivity, responsible for allergic reactions. Parasitic infestations such as hookworm, and certain clinical disorders including aspergillosis, have also been demonstrated to cause high levels of IgE.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty