Would you like to log in again?
DRG® Anti-Phosopholipid Screen IgG/IgM ELISA, DRG International
Supplier: DRG International
DRG®
EIA-3591R
75871-214KT 564.71 USD InStock
75871-214
DRG® Anti-Phosopholipid Screen IgG/IgM ELISA, DRG International
Assays ELISAs
An enzyme immunoassay test system to screen for the presence of IgG and IgM class autoantibodies against cardiolipin, phosphatidyl serine, phosphatidyl inositol, phosphatidic acid and beta-2-glycoprotein I in human serum or plasma.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
A mixture of highly purified cardiolipin, phosphatidyl serine, phosphatidyl inositol, phosphatidic acid and human beta-2-Glycoprotein I is bound to microwells
In primary APS autoantibodies against phospholipids appear independently, while in secondary APS phospholipid antibodies are detected in conjunction with other autoimmune diseases, such as lupus erythematosus, rheumatoid arthritis, or Sjogren’s syndrome. Phospholipid antibodies are detectable in only 1-5 % of healthy individuals, but they are found in 16-35 % of lupus patients. The presence of anti-cardiolipin antibodies in systemic lupus erythematosus (SLE) can be related to the development of thrombosis and thrombocytopenia. In gynecology they are supposed to cause intrauterine death or recurrent abortion. Furthermore, anti-cardiolipin antibodies have been detected in neurological disorders like cerebrovascular insufficiency, cerebral ischemia, epilepsy or chorea. Anticardiolipin autoantibodies occur in the immunoglobulin classes IgG, IgM or IgA. The determination of IgM antibodies is a valuable indicator in the diagnosis of beginning autoimmune diseases, whereas IgG antibodies are present in progressive stages of manifested autoimmune disorders. The determination of IgA antibodies seems to have a greater importance in the African-Caribbean population. Quantitative measurement of anti-cardiolipin antibodies, especially IgG, shows high specificity in therapy-monitoring of secondary APS related to SLE. Clinical indications for determination of anti-cardiolipin antibodies are: SLE, thrombosis, thrombocytopenia, cerebral ischemia, chorea, epilepsy, recurrent abortion, intrauterine death. The discovery that anti-phospholipid antibodies recognize plasma proteins that are associated with phospholipids rather than binding to the phospholipids themselves has been a major advance in APS research. Several reports indicate that beta-2-glycoprotein I antibodies are clinically relevant. Recent studies suggest the presence of a dominant epitope on the first domain of beta-2-glycoprotein I. In contrast to antibodies recognizing other domains of beta-2-glycoprotein I, anti-domain I antibodies are found to be highly associated with clinical symptoms. Anti-cardiolipin and anti-beta-2-glycoprotein I antibodies are independent risk factors for the occurrence of vascular thrombosis and pregnancy loss. However, patients testing positive for multiple antibody specificities generally have a more severe disease and higher recurrence rates despite treatment. Besides the standardized laboratory assays for detection of anti-cardiolipin antibodies, antibodies directed to beta‑2‑gycoprotein I and LA, defined in the classification criteria, several other autoantibodies have shown to be relevant to APS. Among them are antibodies against negatively-charged phospholipids, like phosphatidyl serine, phosphatidyl inositol and phosphatidic acid (PA). These antigens can improve the clinical sensitivity in patient samples with suspected APS but they will not replace the determination of autoantibodies against cardiolipin or beta-2-glycoprotein I. Autoantibodies that bind to proteins of the coagulation cascade or complexes of these proteins with phospholipids have also been proposed to be relevant for APS. As an example, a test for anti-prothrombin antibodies in conjunction with other parameters may be a good risk marker for thrombosis. Antibodies to Annexin V may also be detectable within the clinical framework of APS with otherwise negative phospholipid antibody results.
The determination is based on an indirect enzyme linked immune reaction with the following steps:Specific antibodies in the patient sample bind to the antigen coated on the surface of the reaction wells. After incubation, a washing step removes unbound and unspecifically bound serum or plasma components. Subsequently added enzyme conjugate binds to the immobilized antibody-antigen-complexes. After incubation, a second washing step removes unbound enzyme conjugate. After addition of substrate solution the bound enzyme conjugate hydrolyses the substrate forming a blue coloured product. Addition of an acid stops the reaction generating a yellow end-product. The intensity of the yellow colour correlates with the concentration of the antibody-antigen-complex and can be measured photometrically at 450 nm.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty