Would you like to log in again?
DRG® PAP ELISA, DRG International
Supplier: DRG International
DRG®
EIA-3763R
75871-220KT 787.14 USD InStock
75871-220
DRG® PAP ELISA, DRG International
Assays ELISAs
An enzyme immunoassay for the quantitative measurement of plasmin-α2-antiplasmin complex (PAP) in plasma.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
The removal of polymerized fibrin from the vascular system by proteolytic degradation (fibrinolysis) is important for maintaining the haemostatic balance
The DRG PAP micro ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. During the first incubation step the PAP present in the sample binds to the monoclonal antibodies (PAP-6) coated on the surface of the microtitration plate. Unbound fragments are removed by washing and, in a second reaction, peroxidase-conjugated antibodies to plasminogen are bound to the free PAP determinants. After incubation the unbound conjugate is washed off. The amount of bound peroxidase is proportional to the concentration of PAP in the sample. Having added the substrate solution, the intensity of colour developed is proportional to the concentration of PAP in the patient sample.
The key enzyme of the fibrinolytic system is plasmin. Besides its fibrinolytic function in the plasma, plasmin also plays a central role in the activation of degenerative (e. g. collagenases) and inflammatory (e. g. complement system) processes in the tissue. Defects or deficiencies in a component of the fibrinolytic system can lead to thromboembolic diseases. Conversely, hyperfibrinolytic states increase the risk of haemorrhage through accelerated degradation of the fibrinogen. Plasmin is effectively bound in the plasma by its inhibitor α2-antiplasmin, forming the plasmin‑α2-antiplasmin complex (PAP). The plasma PAP concentration is thus a measure of the current activity of the fibrinolytic system. Elevated PAP concentrations have been reported in patients with DIC. An increase in PAP concentrations has also been observed during fibrinolytic therapies. Due to the complement and collagenase activating functions of plasmin, PAP has also been found to be increased in patients with inflammatory rheumatic diseases.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty