Would you like to log in again?
DRG® FSH ELISA, DRG International, Inc.
Supplier: DRG International
DRG®
EIA-1288
75871-248KT 154.02 USD InStock
75871-248
DRG® FSH ELISA, DRG International, Inc.
Assays ELISAs
An enzyme immunoassay for the quantitative measurement of follicle stimulating hormone (FSH) in serum.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
Follicle-Stimulating Hormone (FSH) and Luteinizing Hormone (LH) are intimately involved in the control of the growth and reproductive activities of the gonadal tissues, which synthesize and secrete male and female sex hormones through a negative feedback relationship
In the female, FSH stimulates the growth and maturation of ovarian follicles by acting directly on the receptors located on the granulosa cells; follicular teroidogenesis is promoted and LH production is stimulated. The LH produced then binds to the theca cells and stimulates steroidogenesis. Increased intraovarian estradiol production occurs as follicular maturation advances, thereupon stimulating increased FSH receptor activity and FSH follicular binding. FSH, LH, and estradiol are therefore intimately related in supporting ovarian recruitment and maturation in women. FSH levels are elevated after menopause, castration, and in premature ovarian failure. The levels of FSH may be normalized through the administration of estrogens, which demonstrate a negative feedback mechanism. Abnormal relationships between FSH and LH, between FSH and estrogen have been linked to anorexia nervosa and polycystic ovarian disease. Although there are significant exceptions ovarian failure is indicated when random FSH concentrations exceed 40 mIU/mL.The growth of the seminiferous tubules and maintenance of spermatogenesis in men are regulated by FSH. However, androgens, unlike estrogens, do not lower FSH levels, therefore demonstrating a feedback relationship only with serum LH. For reasons not fully understood, azospermic and oligospermic males usually have elevated FSH levels. Tumors of the testes generally depress serum FSH concentrations, but levels of LH are elevated, as determined by radioimmunoassay. It has been postulated that the apparent LH increase may be caused by cross reactivity with hCG-like substances secreted by tumors of the testes. High levels of FSH in men may be found in primary testicular failure and Klinefelter syndrome. Elevated concentrations are also present in cases of starvation, renal failure, hyperthyroidism, and cirrhosis.
The DRG FSH ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal antibody directed towards a unique antigenic site on a FSH molecule. An aliquot of sample containing endogenous FSH is incubated in the coated well with enzyme conjugate, which is an anti-FSH monoclonal antibody conjugated with horseradish peroxidase. After incubation the unbound conjugate is washed off. The amount of bound peroxidase is proportional to the concentration of FSH in the sample. Having added the substrate solution, the intensity of color developed is proportional to the concentration of FSH in the sample.
FSH is a glycoprotein secreted by the basophil cells of the anterior pituitary. Gonadotropin-releasing hormone (GnRH), produced in the hypothalamus, controls the release of FSH from the anterior pituitary. Like other glycoproteins, such as LH, TSH, and HCG, FSH consists of subunits designated as alpha and beta. Hormones of this type have alpha subunits that are very similar structurally, therefore the biological and immunological properties of each are dependent on the unique beta subunit.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty