Would you like to log in again?
DRG® LH-Urine ELISA, DRG International
Supplier: DRG International
DRG®
EIA-1290
75871-252KT 155.46 USD InStock
75871-252
DRG® LH-Urine ELISA, DRG International
Assays ELISAs
An enzyme immunoassay for the quantitative measurement of luteinizing hormone (LH) in urine.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
Luteinizing hormone (LH) is produced in both men and women from the anterior pituitary gland in response to luteinizing hormone-releasing hormone (LH-RH or Gn-RH), which is released by the hypothalamus
The DRG LH-Urine ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal antibody directed towards a unique antigenic site of the β‑LH molecule. An aliquot of sample containing endogenous LH is incubated in the coated well with enzyme conjugate, which is an anti- LH antibody conjugated with horseradish peroxidase. After incubation the unbound conjugate is washed off. The amount of bound peroxidase is proportional to the concentration of LH in the sample. Having added the substrate solution, the intensity of color developed is proportional to the concentration of LH in the sample.
LH, also called interstitial cell-stimulating hormone (ICSH) in men, is a glycoprotein with a molecular weight of approximately 30.000 daltons. It is composed of two non covalently associated dissimilar amino acid chains, alpha and beta. The alpha chain is similar to that found in human thyroid-stimulating hormone (TSH), follicle stimulating hormone (FSH), and human chorionic gonadotropin (hCG). The difference between these hormones lie in the amino acid composition of their beta subunits, which account for their immunological differentiation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty