Would you like to log in again?
DRG® Prolactin ELISA, DRG International
Supplier: DRG International
DRG®
EIA-1291
75871-254KT 156.21 USD InStock
75871-254
DRG® Prolactin ELISA, DRG International
Assays ELISAs
An enzyme immunoassay for the quantitative measurement of prolactin in serum.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
Human prolactin (lactogenic hormone) is secreted from the anterior pituitary gland in both men and women
The DRG Prolactin ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal [mouse] antibody directed towards a unique antigenic site on a Prolactin molecule. An aliquot of sample containing endogenous Prolactin is incubated in the coated well with enzyme conjugate, which is an anti- Prolactin antibody conjugated with horseradish peroxidase. After incubation the unbound conjugate is washed off. The amount of bound peroxidase is proportional to the concentration of Prolactin in the sample. Having added the substrate solution, the intensity of color developed is proportional to the concentration of Prolactin in the sample.
Human prolactin is a single chain polypeptide hormone with a molecular weight of approximately 23.000 daltons. The release and synthesis of prolactin is under neuroendocrinal control, primarily through Prolactin Releasing Factor and Prolactin Inhibiting Factor. Women normally have slightly higher basal prolactin levels than men; apparently, there is an estrogen- related rise at puberty and a corresponding decrease at menopause. The primary functions of prolactin are to initiate breast development and to maintain lactation. Prolactin also suppresses gonadal function. During pregnancy, prolactin levels increase progressively to between 10 and 20 times normal values, declining to non-pregnant levels by 3-4 weeks post- partum. Breast feeding mothers maintain high levels of prolactin, and it may take several months for serum concentrations to return to non-pregnant levels. The determination of prolactin concentration is helpful in diagnosing hypothalamic- pituitary disorders. Microadenomas (small pituitary tumors) may cause hyperprolactinemia, which is sometimes associated with male impotence. High prolactin levels are commonly associated with galactorrhea and amenorrhea. Prolactin concentrations have been shown to be increased by estrogens, thyrotropin- releasing hormone (TRH), and several drugs affecting dopaminergic mechanisms. Prolactin levels are elevated in renal disease and hypothyroidism, and in some situations of stress, exercise, and hypoglycemia. Additionally, the release of prolactin is episodic and demonstrates diurnal variation. Mildly elevated prolactin concentrations should be evaluated taking these considerations into account. Prolactin concentrations may also be increased by drugs such as chloropromazine and reserpine, and may be lowered by bromocyptine and L-dopa.The DRG Prolactin ELISA provides a rapid, sensitive, and a reliable assay. The antibodies developed for the test will determine a minimal concentration of human prolactin of 0.35 ng/mL. There is no cross-reactivity with hCG, TSH, LH, FSH, or hGH.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty