Would you like to log in again?
DRG® Candida albicans IgM ELISA, DRG International, Inc.
Supplier: DRG International
DRG®
EIA-3459R
75871-264KT 156.86 USD InStock
75871-264
DRG® Candida albicans IgM ELISA, DRG International, Inc.
Assays ELISAs
An enzyme immunoassay for the qualitative and semiquantitative determination of IgM-class antibodies to Candida albicans in serum.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
Species of Candida are non-mycelia-producing non-ascospore-forming yeast-like fungi which appear as small (4 to 6 µm), oval, thin-walled gram-positive budding cells. The most important representative, Candida albicans, is a facultative pathogen for man.C. albicansis ubiquitous and commonly found as transient flora on normal mucous membranes. Although not pathogenic in healthy humans the fungus may be opportunistic in those suffering from a variety of disorders, and in those treated intensively with broad-spectrum antibiotics or immunosuppressive measures. Candidiasis is caused about 90% of the time byC. albicans. It is an acute or subacute infection in which the fungus may produce lesions in the mouth (thrush, oral candidiasis), vagina (vulvovaginal candidiasis), skin and nails (intertriginous candidiasis), bronchi or lungs (bronchopulmonary candidiasis) and, occasionally, a septicemia, endocarditis or meningitis. In immunosuppressant patients with cellular immunodeficiency, e.g., AIDS patients,C. albicansmay lead to severe necrosis of infected tissues. The DRG Candida albicans IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) Microtiter wells as a solid phase are coated with Candida albicans antigen. Diluted patient specimens and ready-for-use controls are pipetted into these wells. During incubation Candida albicans-specific antibodies of positive specimens and controls are bound to the immobilized antigens. After a washing step to remove unbound sample and control material horseradish peroxidase conjugated anti-human IgG antibodies are dispensed into the wells. During a second incubation this anti‑IgG conjugate binds specifically to IgG antibodies resulting in the formation of enzyme-linked immune complexes. After a second washing step to remove unbound conjugate the immune complexes formed (in case of positive results) are detected by incubation with TMB substrate and development of a blue color. The blue color turns into yellow by stopping the enzymatic indicator reaction with sulfuric acid. The intensity of this color is directly proportional to the amount of Candida albicans-specific IgG antibody in the patient specimen. Absorbance at 450 nm is read using an ELISA microtiter plate reader.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty