Would you like to log in again?
DRG® Free Estriol ELISA, DRG International, Inc.
Supplier: DRG International
DRG®
EIA-1612
75871-280KT 152.6 USD InStock
75871-280
DRG® Free Estriol ELISA, DRG International, Inc.
Assays ELISAs
An enzyme immunoassay for the quantitative measurement of free estriol (unconjugated estriol) in serum during the second half of pregnancy.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
Estriol (E3) is the major estrogen formed by the fetoplacental unit during pregnancy
The DRG Free Estriol ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. The microtiter wells are coated with a polyclonal [rabbit] antibody directed towards an antigenic site on the Estriol molecule. Endogenous Estriol of a sample competes with an Estriol-horseradish peroxidase conjugate for binding to the coated antibody. After incubation the unbound conjugate is washed off. The amount of bound peroxidase conjugate is inversely proportional to the concentration of Estriol in the sample. After addition of the substrate solution, the intensity of color developed is inversely proportional to the concentration of Estriol in the sample.
Unconjugated E3 passes through the placenta into the maternal circulation, where it is rapidly converted into glucuronide and sulfate derivatives to facilitate its excretion. The half-life of estriol in the maternal bloodstream is only 20 to 30 minutes. Its measurement, therefore offers a convenient and quick evaluation of current fetal status. Plasma estriol levels increase steadily throughout pregnancy and most rapidly during the third trimester (28 to 40 weeks). A sudden decrease in fetoplacental E3 production will result in a rapid fall in unconjugated E3 in the maternal serum. There are several potential advantages to measuring unconjugated E3 rather than total serum or urinary E3. Unconjugated estriol levels are free from effects related to maternal renal or hepatic disease, and are not altered by the administration of certain antibiotics. Unconjugated E3 more accurately reflects fetal outcome in diabetic pregnancies. Since no hydrolysis of unconjugated E3 is required, a more rapid tumaround for the test result is possible.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty