Would you like to log in again?
DRG® Insulin ELISA, DRG International, Inc.
Supplier: DRG International
DRG®
EIA-2935
75871-320KT 242.41 USD InStock
75871-320
DRG® Insulin ELISA, DRG International, Inc.
Assays ELISAs
An enzyme immunoassay for the quantitative measurement of insulin in serum and plasma.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
Insulin is the principal hormone responsible for the control of glucose metabolism
The DRG Insulin ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal antibody directed towards a unique antigenic site on the Insulin molecule. An aliquot of patient sample containing endogenous Insulin is incubated in the coated well with enzyme conjugate, which is an anti-Insulin antibody conjugated with Biotin. After incubation the unbound conjugate is washed off. During the second incubation step Streptavidin Peroxidase Enzyme Complex binds to the biotin-anti-Insulin antibody. The amount of bound HRP complex is proportional to the concentration of Insulin in the sample. Having added the substrate solution, the intensity of colour developed is proportional to the concentration of Insulin in the patient sample.
It is synthesized in the β‑cells of the islets of Langerhans as the precursor, proinsulin, which is processed to form C-peptide and insulin. Both are secreted in equimolar amounts into the portal circulation. The mature insulin molecule comprises two polypeptide chains, the A chain and B chain (21 and 30 amino acids respectively). The two chains are linked together by two inter-chain disulphide bridges. There is also an intra-chain disulphide bridge in the A chain. Secretion of insulin is mainly controlled by plasma glucose concentration, and the hormone has a number of important metabolic actions. Its principal function is to control the uptake and utilisation of glucose in peripheral tissues via the glucose transporter. This and other hypoglycaemic activities, such as the inhibition of hepatic gluconeogenesis and glycogenolysis are counteracted by the hyperglycaemic hormones including glucagon, epinephrine (adrenaline), growth hormone and cortisol. Insulin concentrations are severely reduced in insulin-dependent diabetes mellitus (IDDM) and some other conditions such as hypopituitarism. Insulin levels are raised in non-insulin-dependent diabetes mellitus (NIDDM), obesity, insulinoma and some endocrine dysfunctions such as Cushing’s syndrome and acromegaly.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty