Would you like to log in again?
DRG® Androstenedione ELISA, DRG International, Inc.
Supplier: DRG International
DRG®
EIA-3265
75871-324KT 242.41 USD InStock
75871-324
DRG® Androstenedione ELISA, DRG International, Inc.
Assays ELISAs
An enzyme immunoassay for the quantitative measurement of androstenedione in serum and plasma.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
The steroid hormone Androstenedione is one of the main androgens, besides Testosterone and Dehydroepiandrosterone
The DRG Androstenedione ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. The microtiter wells are coated with a polyclonal [rabbit] antibody directed towards an antigenic site of the Androstenedione molecule. Endogenous Androstenedione of a patient sample competes with an Androstenedione-horseradish peroxidase conjugate for binding to the coated antibody. After incubation the unbound conjugate is washed off. The amount of bound peroxidase conjugate is inversely proportional to the concentration of Androstenedione in the sample. After addition of the substrate solution, the intensity of colour developed is inversely proportional to the concentration of Androstenedione in the patient sample.
Testosterone, the most important biological active androgen, is derived from peripheral enzymatic conversion of Androstenedione. In males, androgens are secreted primarily by the Leydig cells of the testes, to some degree also in the adrenal cortex. In females, the androgens are secreted mainly in the adrenal glands and in the ovary. Around 10% of the androgens are derived from peripheral conversion, mainly of DHEA. Androstenedione and Testosterone show high diurnal variability. The highest levels are measured in the morning. At the age of puberty serum androstenedione levels rise, after menopause they decline again. High androstenedione levels are measured during pregnancy. In women, high levels of androstenedione (47-100% above normal) are generally found in hirsutism, mostly in combination with other androgens as testosterone and DHEA-S. Androstenedione overproduction is due to ovarian dysfunction or may be of adrenal origin. High circulating androstenedione levels are found in women with polycystic ovaries and 21-hydroxylase effect. Significant lower androstenedione levels are found in postmenopausal osteoporosis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty