Would you like to log in again?
DRG® Myoglobin ELISA, DRG International
Supplier: DRG International
DRG®
EIA-3955
75871-358KT 264.38 USD InStock
75871-358
DRG® Myoglobin ELISA, DRG International
Assays ELISAs
An enzyme immunoassay for the quantitative determination of myoglobin concentration in serum.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
Myoglobin, a heme protein with a molecular weight of approximately 17,500 Daltons is found in both cardiac and skeletal muscle
A number of reports suggest using the measurement of myoglobin as a diagnostic aid in ruling out myocardial infarction with negative predictive values of up to 100% reported at certain time periods after the onset of symptoms. Unlike the other cardiac enzymes such as creeatine kinase and the MB isoform (i.e., CK and CK/MB) which do not reach serum levels until several hours post-infarction (approx. 19 hours), myoglobin levels can be expected to peak within 6 to 9 hours. The Myoglobin Enzyme Immunoassay provides a rapid, sensitive, and reliable assay for the quantitative measurement of myoglobin in serum. The antibodies developed for the test will determine a minimal concentration of 5.0 ng/ml, and there is no cross-reactivity with related cardiac or skeletal enzymes. The Myoglobin ELISA test is based on the principle of a solid phase enzyme-linked immunosorbent assay. The assay system utilizes a unique monoclonal antibody directed against a distinct antigenic determinant on the myoglobin molecule. Mouse monoclonal anti-myoglobin antibody is used for solid phase immobilization (on the microtiter wells). A goat anti-myoglobin antibody is in the antibody-enzyme (horseradish peroxidase) conjugate solution. The test sample is allowed to react simultaneously with the two antibodies, resulting in the myoglobin molecules being sandwiched between the solid phase and enzyme-linked antibodies. After a 45-minute incubation at room temperature, the wells are washed with water to remove unbound labeled antibodies. A TMB (Tetramethyl-benzidine) Reagent is added and incubated for 20 minutes, resulting in the development of a blue color. The color development is stopped with the addition of Stop Solution changing the color to yellow. The concentration of myoglobin is directly proportional to the color intensity of the test sample. Absorbance is measured spectrophotometrically at 450 nm.
Damage to either type of muscle following conditions such as trauma, ischemia, and diseases that cause myopathy, is associated with the release of myoglobin into serum. Specifically, following cardiac necrosis associated with myocardial infarction (MI), myoglobin is one of the first markers to rise above normal levels. Myoglobin levels increase measurably above baseline within 2 to 4 hours post-infarct, peaking at 9 to 12 hours, and returning to baseline within 24 to 36 hours. In the absence of skeletal muscle trauma or other factors associated with a non-cardiac related increase in circulating myoglobin, its levels have been used as an early marker for myocardial infarct.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty