Would you like to log in again?
DRG® HSV-1+2 IgM ELISA, DRG International, Inc.
Supplier: DRG International
DRG®
EIA-3490R
75871-378KT 259.54 USD InStock
75871-378
DRG® HSV-1+2 IgM ELISA, DRG International, Inc.
Assays ELISAs
An enzyme immunoassay for the qualitative and semiquantitative determination of IgM-class antibodies to herpes simplex virus type 1 + 2 in human serum and plasma.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
Herpes simplex is an enveloped DNA virus (150 to 200 nm in diameter) belonging to the alphaherpesviridae
The DRG Herpes Simplex Virus Type 1+2 IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) Microtiter wells as a solid phase are coated with recombinant gG1 Protein of HSV-1 and native gG2 protein of HSV-2. Diluted patient specimens and ready-for-use controls are pipetted into these wells. During incubation Herpes Simplex Virus Type 1+2-specific antibodies of positive specimens and controls are bound to the immobilized antigens. After a washing step to remove unbound sample and control material horseradish peroxidase conjugated anti-human IgG antibodies are dispensed into the wells. During a second incubation this anti‑IgG conjugate binds specifically to IgG antibodies resulting in the formation of enzyme-linked immune complexes. After a second washing step to remove unbound conjugate the immune complexes formed (in case of positive results) are detected by incubation with TMB substrate and development of a blue color. The blue color turns into yellow by stopping the enzymatic indicator reaction with sulfuric acid. The intensity of this color is directly proportional to the amount of Herpes Simplex Virus Type 1+2-specific IgG antibody in the patient specimen. Absorbance at 450 nm is read using an ELISA microtiter plate reader.
Based on antigenic, biochemical and biological differences it can be divided into two serotypes, HSV-1 and HSV-2. Man is the only known natural host and source of the virus. HSV-type 1 typically causes oral herpes, while HSV-type 2 typically affects the genital area. Most of the time, HSV-1 and HSV-2 are inactive, or “silent”, and cause no symptoms, but some infected people have “outbreaks” of blisters and ulcers. Once infected with HSV, people remain infected for life. Herpes simplex viruses are among the most common infectious agents of man, and either HSV type appears to be capable of infecting similar body sites. A high percentage of the adult population is seropositive (approx. 90% HSV-1 is dependent on socio-economic status, 10 to 30% HSV-2). Primary HSV-1 infection usually occurs in early childhood (6 to 18 months of age). HSV-2 usually produces mild symptoms, and most people have no recognized symptoms.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty