Would you like to log in again?
DRG® Salivary Testosterone ELISA, DRG International, Inc.
Supplier: DRG International
DRG®
SLV-3013
75871-386KT 213.01 USD InStock
75871-386
DRG® Salivary Testosterone ELISA, DRG International, Inc.
Assays ELISAs
An enzyme immunoassay for the quantitative measurement of free active testosterone in saliva.
  • FDA cleared tests
  • Increased stability and linearity
  • Ready-to-use reagents and user-friendly assays
  • Measurement of the free (unbound) fraction of steroids
  • Excellent correlation to mass spectrometry
Testosterone (17β-hydroxy-4-androstene-3-one) is a C19 steroid with an unsaturated bond between C-4 and C-5, a ketone group in C-3 and a hydroxyl group in the β position at C-17
Testosterone is responsible for the development of secondary male sex characteristics and its measurements are helpful in evaluating the hypogonadal status. In women high levels of testosterone are generally found in hirsutism and virilisation, polycystic ovaries, ovarian tumors, adrenal tumors and adrenal hyperplasia. In men high levels of testosterone are associated with hypothalamic-pituitary-unit dysfunction, testicular tumors, congenital adrenal hyperplasia and prostate cancer. Low levels of testosterone are encountered in male patients with the following diseases: Klinefelter´s syndrome. Hypopituitarism, testicular feminization, orchidectomy, cryptorchidism, enzymatic defects and some autoimmune diseases.
The DRG Salivary Testosterone ELISA Kit is based on the competition principle and the microplate separation. An unknown amount of free testosterone present in the sample and a fixed amount of testosterone conjugated with horseradish peroxidase compete for the binding sites of mouse monoclonal testosterone antiserum coated onto the wells. After one-hour incubation the microplate is washed to stop the competition reaction. After addition of the substrate solution the concentration of testosterone is inversely proportional to the optical density measured.
This steroid hormone has a molecular weight of 288.4 daltons. Testosterone is the most important androgen secreted into the blood. In males, primarily the Leydig cells of the testis secrete testosterone; in females approximately 50% of circulating testosterone is derived from peripheral conversion of androstenedione, approximately 25% from the ovary and 25% from the adrenal glands.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty